Assay Details
Target Gene Details
Entrez Gene ID: | 21916 |
Gene Name: | tropomodulin 1 |
Gene Aliases: |
E-Tmod, Tmod |
Location: |
Chr.4:46038940-46116032 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmod1 | NM_021883.2 | NP_068683.1 | ||
| AK132878.1 | ||||
| BC106849.1 | AAI06850.1 | |||
| BC106850.1 | AAI06851.1 | |||
| BY127972.1 | ||||
| S76831.1 | AAB33388.1 |
More Information
Set Membership: |
|

Genomic Map