Assay Details
Target Gene Details
Entrez Gene ID: | 56615 |
Gene Name: | microsomal glutathione S-transferase 1 |
Gene Aliases: |
1500002K10Rik, Gst |
Location: |
Chr.6:138140537-138156755 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgst1 | NM_019946.4 | NP_064330.2 | ||
| AF159050.1 | AAD51096.1 | |||
| AK002800.1 | BAB22368.1 | |||
| AK005122.1 | BAB23827.1 | |||
| BC009155.1 | AAH09155.1 |
More Information
Set Membership: |
|

Genomic Map