Assay Details
Target Gene Details
Entrez Gene ID: | 237336 |
Gene Name: | TATA box binding protein-like 1 |
Gene Aliases: |
4732475G08, AW011832, AW491032, D18347, STD, TLF, TRF2, TRP, Tlp |
Location: |
Chr.10:22703877-22732573 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbpl1 | NM_011603.5 | NP_035733.1 | ||
| AB017697.1 | BAA74578.1 | |||
| AK005604.1 | ||||
| AK049975.1 | BAC34014.1 | |||
| AK078547.1 | BAC37330.1 | |||
| BC033926.1 | AAH33926.1 |
More Information
Set Membership: |
|

Genomic Map