Assay Details
Target Gene Details
Entrez Gene ID: | 58233 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 4 |
Gene Aliases: |
1110021L12Rik, AV358213, Dj4, Hsj4, mmDjA4 |
Location: |
Chr.9:54696590-54716317 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja4 | NM_021422.4 | NP_067397.1 | ||
| AB032401.1 | BAA92775.1 | |||
| AK046476.1 | BAC32747.1 | |||
| AK076175.1 | BAC36232.1 | |||
| AK141909.1 | BAE24880.1 | |||
| AK197546.1 | ||||
| BC147484.1 | ||||
| BC147486.1 |
More Information
Set Membership: |
|

Genomic Map