Assay Details
Target Gene Details
Entrez Gene ID: | 69553 |
Gene Name: | family with sequence similarity 65, member C |
Gene Aliases: |
2310033K02Rik, Gm124 |
Location: |
Chr.2:167980161-168050887 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam65c | NM_001080708.2 | NP_001074177.1 | ||
| AK158217.1 | ||||
| BC130272.1 |
More Information
Set Membership: |
|

Genomic Map