Assay Details
Target Gene Details
Entrez Gene ID: | 19899 |
Gene Name: | ribosomal protein L18 |
Gene Aliases: |
L18, Rpl18a |
Location: |
Chr.7:45718071-45720835 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl18 | NM_009077.2 | NP_033103.2 | ||
| AK007263.1 | BAB24923.1 | |||
| AK009044.1 | BAB26043.1 | |||
| AK010510.1 | BAB26993.1 | |||
| AK012580.1 | BAB28332.1 | |||
| AK145649.1 | BAE26565.1 | |||
| AK151667.1 | BAE30593.1 | |||
| AK153172.1 | BAE31777.1 | |||
| AK166803.1 | BAE39031.1 | |||
| AK168288.1 | BAE40233.1 | |||
| AK169659.1 | BAE41282.1 | |||
| BC081468.1 | AAH81468.1 | |||
| BC082290.1 | AAH82290.1 | |||
| BC091732.1 | AAH91732.1 | |||
| DQ403025.1 | ||||
| L04128.1 | AAA40067.1 |
More Information
Set Membership: |
|

Genomic Map