Assay Details
Target Gene Details
Entrez Gene ID: | 228770 |
Gene Name: | R-spondin family, member 4 |
Gene Aliases: |
A730099F22, A930029K19Rik |
Location: |
Chr.2:151842579-151874668 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rspo4 | NM_001040689.1 | NP_001035779.1 | ||
| AK020904.1 | BAC25643.1 | |||
| AK043484.1 | ||||
| BC048707.1 | ||||
| BC116367.1 |
More Information
Set Membership: |
|

Genomic Map