Assay Details
Target Gene Details
Entrez Gene ID: | 56699 |
Gene Name: | CDC42 effector protein (Rho GTPase binding) 4 |
Gene Aliases: |
1500041M20Rik, Borg4, CEP4 |
Location: |
Chr.11:113726850-113752543 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42ep4 | NM_001163346.1 | NP_001156818.1 | ||
| NM_020006.2 | NP_064390.1 | |||
| AB035088.1 | BAA95932.1 | |||
| AF165114.1 | AAD47822.1 | |||
| AK075739.1 | BAC35920.1 | |||
| AK146141.1 | BAE26930.1 | |||
| AK165318.1 | BAE38136.1 | |||
| AK168364.1 | BAE40299.1 | |||
| BC003857.1 | AAH03857.1 | |||
| BM950172.1 |
More Information
Set Membership: |
|

Genomic Map