Assay Details
Target Gene Details
Entrez Gene ID: | 75458 |
Gene Name: | chemokine-like factor |
Gene Aliases: |
1700001C14Rik, 1810018M11Rik, C32, CKLF1, CKLF3, CKLF4, CKLF5, Cklf2, Cklf6, HSPC224, UCK-1 |
Location: |
Chr.8:104250861-104264936 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cklf | NM_001037841.3 | NP_001032930.1 | ||
| NM_001286383.1 | NP_001273312.1 | |||
| NM_029295.3 | NP_083571.1 | |||
| NR_104433.1 | ||||
| AF401531.2 | AAK94043.2 | |||
| AK005577.1 | BAB24133.1 | |||
| AK032759.1 | ||||
| AK046397.1 | BAC32704.1 | |||
| AK050112.1 | BAC34072.1 | |||
| AK086063.1 | BAC39602.1 | |||
| AY046597.1 | AAL05945.1 | |||
| AY047360.1 | AAL10703.1 |
More Information
Set Membership: |
|

Genomic Map