Assay Details
Target Gene Details
Entrez Gene ID: | 75040 |
Gene Name: | EF-hand calcium binding domain 10 |
Gene Aliases: |
4930504H06Rik |
Location: |
Chr.12:33394854-33401269 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab10 | NM_029152.1 | NP_083428.1 | ||
| AK015697.1 | BAB29935.1 | |||
| BF459438.1 |
More Information
Set Membership: |
|

Genomic Map