Assay Details
Target Gene Details
Entrez Gene ID: | 76389 |
Gene Name: | ankyrin repeat domain 36 |
Gene Aliases: |
1700008J08Rik, 1700012M14Rik, Ankrd48 |
Location: |
Chr.11:5569680-5693796 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd36 | NM_023816.2 | NP_076305.2 | ||
| AF294328.1 | AAG45966.1 | |||
| AK005925.1 | BAB24320.1 | |||
| AK132751.1 | ||||
| AK134466.1 | ||||
| BC120815.1 | ||||
| BC120817.1 |
More Information
Set Membership: |
|

Genomic Map