Assay Details
Target Gene Details
Entrez Gene ID: | 242316 |
Gene Name: | growth differentiation factor 6 |
Gene Aliases: |
BMP-13, BMP13, GDF16 |
Location: |
Chr.4:9844372-9862345 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf6 | NM_013526.1 | NP_038554.1 | ||
| AJ537425.1 | CAD60935.1 | |||
| BC141339.1 | ||||
| BC141340.1 |
More Information
Set Membership: |
|

Genomic Map