Assay Details
Target Gene Details
Entrez Gene ID: | 11858 |
Gene Name: | Rho family GTPase 2 |
Gene Aliases: |
Arhn, Rohn |
Location: |
Chr.11:101468338-101471306 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnd2 | NM_009708.1 | NP_033838.1 | ||
| AF016482.1 | AAB68844.1 | |||
| BC127034.1 | ||||
| BC128475.1 | ||||
| BC139330.1 | ||||
| BC139331.1 |
More Information
Set Membership: |
|

Genomic Map