Assay Details
Target Gene Details
Entrez Gene ID: | 20888 |
Gene Name: | sulfotransferase family, cytosolic, 1C, member 1 |
Gene Aliases: |
(PST)G, P-SULT, ST1C1, Stp2, Sult1a2, mOLFST |
Location: |
Chr.17:53961615-53990631 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult1c1 | NM_018751.2 | NP_061221.2 | ||
| AF033653.1 | AAC17740.1 | |||
| BC045149.1 | AAH45149.1 |
More Information
Set Membership: |
|

Genomic Map