Assay Details
Target Gene Details
Entrez Gene ID: | 67864 |
Gene Name: | Yip1 domain family, member 4 |
Gene Aliases: |
2310034L04Rik |
Location: |
Chr.17:74489493-74500277 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf4 | NM_026417.4 | NP_080693.2 | ||
| AK009618.1 | ||||
| AK017721.1 | ||||
| AK017722.1 | ||||
| AK083301.1 | BAC38852.1 | |||
| AK159227.1 | BAE34913.1 | |||
| AK166347.1 | BAE38721.1 | |||
| BC003936.1 | AAH03936.1 | |||
| BY709789.1 |
More Information
Set Membership: |
|

Genomic Map