Assay Details
Target Gene Details
Entrez Gene ID: | 27362 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 9 |
Gene Aliases: |
AA408011, AA673251, AA673481, AW556981, ERdj4, Mdg1, mDj7 |
Location: |
Chr.12:44205897-44210068 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb9 | NM_013760.4 | NP_038788.2 | ||
| AB028857.1 | BAA88305.1 | |||
| AK005475.1 | BAB24065.1 | |||
| BC042713.1 | AAH42713.1 | |||
| BC096676.1 | AAH96676.1 | |||
| CT010304.1 | CAJ18512.1 |
More Information
Set Membership: |
|

Genomic Map