Assay Details
Target Gene Details
Entrez Gene ID: | 77090 |
Gene Name: | occludin/ELL domain containing 1 |
Gene Aliases: |
9430098E02Rik, AI326022, AI593544 |
Location: |
Chr.8:71371298-71373689 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ocel1 | NM_001293800.1 | NP_001280729.1 | ||
| NM_029865.2 | NP_084141.2 | |||
| NR_122030.1 | ||||
| AK020528.1 | BAB32126.1 | |||
| AK155219.1 | BAE33130.1 | |||
| BB766991.1 | ||||
| BC019407.1 | AAH19407.1 |
More Information
Set Membership: |
|

Genomic Map