Assay Details
Target Gene Details
Entrez Gene ID: | 68026 |
Gene Name: | RIKEN cDNA 2810417H13 gene |
Gene Aliases: |
AA409629, Ns5apt9, Ns5atp9, PAF15, Paf, mKIAA0101, p15(PAF) |
Location: |
Chr.9:65890300-65903553 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2810417H13Rik | NM_026515.2 | NP_080791.2 | ||
| AK011090.1 | BAB27391.1 | |||
| AK011645.1 | ||||
| AK011835.1 | BAB27868.1 | |||
| AK012140.1 | ||||
| AK013105.1 | BAB28650.1 | |||
| AK017673.1 | BAB30866.1 | |||
| AK134432.1 | 2 | 2259 | ||
| AK212319.1 | ||||
| AY496943.1 | AAR90858.1 | |||
| BC021413.1 | AAH21413.1 | |||
| BC096393.1 | AAH96393.1 |
Target Gene Details
Entrez Gene ID: | 56404 |
Gene Name: | thyroid hormone receptor interactor 4 |
Gene Aliases: |
4930558E03Rik, ASC-1, Asc1, BB191711 |
Location: |
Chr.9:65828926-65908809 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip4 | NM_019797.4 | NP_062771.2 | ||
| AF539614.1 | AAN23117.1 | |||
| BB617447.1 |
More Information
Set Membership: |
|

Genomic Map