Assay Details
Target Gene Details
Entrez Gene ID: | 225030 |
Gene Name: | potassium voltage-gated channel, subfamily G, member 3 |
Gene Aliases: |
KV6.3, Kv10.1a, Kv10.1b |
Location: |
Chr.17:83585411-83631993 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcng3 | NM_153512.1 | NP_705732.1 | ||
| AF454551.1 | AAM93552.1 | |||
| AF454552.1 | AAM93553.1 |
More Information
Set Membership: |
|

Genomic Map