Assay Details
Target Gene Details
Entrez Gene ID: | 69017 |
Gene Name: | proline-rich transmembrane protein 2 |
Gene Aliases: |
1500031I19Rik, AI195361, DSPB3 |
Location: |
Chr.7:127017542-127021226 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prrt2 | NM_001102563.1 | NP_001096033.1 | ||
| AK158895.1 | ||||
| BB872849.1 | ||||
| BB873187.1 |
More Information
Set Membership: |
|

Genomic Map