Assay Details
Target Gene Details
Entrez Gene ID: | 20044 |
Gene Name: | ribosomal protein S14 |
Gene Aliases: |
2600014J02Rik, AL023078 |
Location: |
Chr.18:60774596-60778546 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps14 | NM_020600.4 | NP_065625.2 | ||
| AK003148.1 | BAB22604.1 | |||
| AK011213.1 | BAB27472.1 | |||
| AK012425.1 | BAB28230.1 | |||
| AK012584.1 | BAB28334.1 | |||
| AK019233.1 | BAB31615.1 | |||
| AK028106.1 | BAC25751.1 | |||
| AK146325.1 | BAE27080.1 | |||
| AK146976.1 | BAE27580.1 | |||
| AK152461.1 | 1 | 1459 | ||
| AK153401.1 | 1 | 1459 | ||
| AK168237.1 | BAE40187.1 | |||
| BY149058.1 |
More Information
Set Membership: |
|

Genomic Map