Assay Details
Target Gene Details
Entrez Gene ID: | 286940 |
Gene Name: | filamin, beta |
Gene Aliases: |
AL024016, Fln-b |
Location: |
Chr.14:7817957-7951588 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flnb | NM_001081427.1 | NP_001074896.1 | ||
| NM_134080.1 | NP_598841.1 | |||
| AK050046.1 | ||||
| BP765788.1 |
More Information
Set Membership: |
|

Genomic Map