Assay Details
Target Gene Details
Entrez Gene ID: | 241324 |
Gene Name: | crumbs family member 2 |
Gene Aliases: |
5930402A21, BC043114 |
Location: |
Chr.2:37776225-37799103 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crb2 | NM_001163566.1 | NP_001157038.1 | ||
| BC043114.1 | AAH43114.1 |
More Information
Set Membership: |
|

Genomic Map