Assay Details
Target Gene Details
Entrez Gene ID: | 622127 |
Gene Name: | cytochrome P450, family 3, subfamily a, polypeptide 57 |
Gene Aliases: |
EG622127 |
Location: |
Chr.5:145345145-145390926 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp3a57 | NM_001100180.1 | NP_001093650.1 |
More Information
Set Membership: |
|

Genomic Map