Assay Details
Target Gene Details
Entrez Gene ID: | 56388 |
Gene Name: | cytochrome P450, family 3, subfamily a, polypeptide 25 |
Gene Aliases: |
AI327008 |
Location: |
Chr.5:145977194-146009639 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp3a25 | NM_019792.2 | NP_062766.2 | ||
| NR_030782.1 | ||||
| AF204959.1 | AAF15303.1 | |||
| AK149541.1 | ||||
| BC028855.1 | AAH28855.1 | |||
| BY099368.1 | ||||
| Y11995.1 | CAA72720.1 |
More Information
Set Membership: |
|

Genomic Map