Assay Details
Target Gene Details
Entrez Gene ID: | 231821 |
Gene Name: | ArfGAP with dual PH domains 1 |
Gene Aliases: |
4930431P11Rik, Centa1, GC1L, p42IP4 |
Location: |
Chr.5:139271876-139325464 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adap1 | NM_172723.4 | NP_766311.2 | ||
| BY740716.1 | ||||
| CX242691.1 |
More Information
Set Membership: |
|

Genomic Map