Assay Details
Target Gene Details
Entrez Gene ID: | 228802 |
Gene Name: | BPI fold containing family B, member 5 |
Gene Aliases: |
Lplunc5 |
Location: |
Chr.2:154223742-154240902 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifb5 | NM_144890.2 | NP_659139.2 | ||
| AK142982.1 | BAE25239.1 | |||
| BC018465.1 | AAH18465.1 |
More Information
Set Membership: |
|

Genomic Map