Assay Details
Target Gene Details
Entrez Gene ID: | 230809 |
Gene Name: | PDLIM1 interacting kinase 1 like |
Gene Aliases: |
BC027088, stk35l2 |
Location: |
Chr.4:134275002-134287846 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdik1l | NM_001163794.1 | NP_001157266.1 | ||
| NM_146156.3 | NP_666268.1 | |||
| AK053750.1 | BAC35506.1 | |||
| AK139413.1 | BAE24001.1 | |||
| AK141566.1 | BAE24738.1 | |||
| AK141823.1 | BAE24845.1 | |||
| AK157013.1 | BAE33930.1 | |||
| AK162350.1 | BAE36868.1 | |||
| AK164975.1 | BAE37986.1 | |||
| BC027088.1 | AAH27088.1 | |||
| BC058514.1 | AAH58514.1 |
More Information
Set Membership: |
|

Genomic Map