Assay Details
Target Gene Details
Entrez Gene ID: | 225895 |
Gene Name: | TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor |
Gene Aliases: |
2810417N14Rik, BB223262, C530024J06Rik, Paf65a |
Location: |
Chr.19:8774354-8786417 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf6l | NM_001177798.1 | NP_001171269.1 | ||
| NM_146092.2 | NP_666204.2 | |||
| AK077149.1 | BAC36645.1 | |||
| AK144881.1 | ||||
| BC028647.1 | AAH28647.1 | |||
| BY295292.1 |
More Information
Set Membership: |
|

Genomic Map