Assay Details
Target Gene Details
Entrez Gene ID: | 66916 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7 |
Gene Aliases: |
1110002H15Rik |
Location: |
Chr.8:83566758-83571623 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb7 | NM_025843.3 | NP_080119.1 | ||
| AK002678.1 | BAB22280.1 | |||
| AK003300.1 | BAB22701.1 | |||
| BC034787.1 | AAH34787.1 |
More Information
Set Membership: |
|

Genomic Map