Assay Details
Target Gene Details
Entrez Gene ID: | 56520 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 4 |
Gene Aliases: |
2610027N22Rik, 2810024O08Rik, 5730493H09Rik, NM23-M4, Nm23M4 |
Location: |
Chr.17:26091734-26095527 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme4 | NM_019731.1 | NP_062705.1 | ||
| AF153451.1 | AAD38977.1 | |||
| AF288690.1 | AAG02200.1 | |||
| AK011573.1 | BAB27708.1 | |||
| AK012805.1 | ||||
| AK017724.1 | BAB30896.1 | |||
| BC027277.1 | AAH27277.1 |
More Information
Set Membership: |
|

Genomic Map