Assay Details
Target Gene Details
Entrez Gene ID: | 56632 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
C76851 |
Location: |
Chr.7:45709463-45718002 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sphk2 | NM_001172561.1 | 6 | 2236 | NP_001166032.1 |
| NM_020011.5 | 6 | 2545 | NP_064395.2 | |
| NM_203280.3 | 7 | 2307 | NP_975009.1 | |
| AF245448.1 | 7 | 2257 | AAF74125.1 | |
| AF415214.1 | AAL07500.1 | |||
| AK004951.1 | 6 | 2545 | BAB23694.1 | |
| AK009037.1 | ||||
| AK016616.1 | 3 | 1325 | ||
| AK146414.1 | 6 | 2138 | ||
| AK151990.1 | 7 | 2207 | BAE30855.1 | |
| AK154321.1 | 6 | 1973 | BAE32512.1 | |
| BC006941.1 | 6 | 1979 | AAH06941.1 | |
| BC053737.1 | 7 | 2220 | AAH53737.1 | |
| BC092084.1 | 6 | 1994 | AAH92084.1 |
More Information
Set Membership: |
|

Genomic Map