Assay Details
Target Gene Details
Entrez Gene ID: | 100141474 |
Gene Name: | RIKEN cDNA 4933428G20 gene |
Gene Aliases: |
- |
Location: |
Chr.11:97476180-97500340 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933428G20Rik | NM_001289118.1 | NP_001276047.1 | ||
| AK016967.1 | BAB30528.1 |
Target Gene Details
Entrez Gene ID: | 58996 |
Gene Name: | Rho GTPase activating protein 23 |
Gene Aliases: |
A330041B18Rik |
Location: |
Chr.11:97404393-97502402 on Build GRCm38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap23 | NM_021493.2 | NP_067468.2 | ||
| AB041572.1 | BAA95056.1 | |||
| AK079135.1 | BAC37556.1 | |||
| AK173187.1 | BAD32465.1 |
More Information
Set Membership: |
|

Genomic Map