Assay Details
Target Gene Details
Entrez Gene ID: | 66477 |
Gene Name: | upregulated during skeletal muscle growth 5 |
Gene Aliases: |
2010301L15Rik |
Location: |
Chr.19:47083471-47090625 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usmg5 | NM_023211.4 | NP_075700.2 | ||
| AK008345.1 | ||||
| AK038457.1 | ||||
| AK088856.1 | ||||
| BC087919.1 | AAH87919.1 |
More Information
Set Membership: |
|

Genomic Map