Assay Details
Target Gene Details
Entrez Gene ID: | 16898 |
Gene Name: | ribosomal protein S2 |
Gene Aliases: |
Llrep3, S4 |
Location: |
Chr.17:24718121-24721927 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Target Gene Details
Entrez Gene ID: | 104366 |
Gene Name: | small nucleolar RNA, H/ACA box 64 |
Gene Aliases: |
MBI-29, Rnu64 |
Location: |
Chr.17:24720789-24720905 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snora64 | NR_002897.1 | |||
| AF357417.1 |
More Information
Set Membership: |
|

Genomic Map