Assay Details
Target Gene Details
Entrez Gene ID: | 448850 |
Gene Name: | zinc finger, HIT type 3 |
Gene Aliases: |
Myohd1, Trip3 |
Location: |
Chr.11:84910950-84916356 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Znhit3 | NM_001005223.2 | 5 | 476 | NP_001005223.1 |
AK002888.1 | 5 | 468 | BAB22434.1 | |
AK003721.1 | 5 | 475 | BAB22958.1 | |
AK004259.1 | 5 | 478 | BAB23239.1 | |
AK014350.1 | 5 | 487 | BAB29291.1 | |
BC089554.1 | 5 | 564 | AAH89554.1 |
