Assay Details
Target Gene Details
Entrez Gene ID: | 17448 |
Gene Name: | malate dehydrogenase 2, NAD (mitochondrial) |
Gene Aliases: |
MDH, Mdh-2, Mor-1, Mor1 |
Location: |
Chr.5:135778649-135790386 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdh2 | NM_008617.2 | NP_032643.2 | ||
| AK002305.1 | BAC24986.1 | |||
| AK135162.1 | BAE22447.1 | |||
| AK160553.1 | BAE35869.1 | |||
| AK167809.1 | BAE39836.1 | |||
| BC023482.1 | AAH23482.1 | |||
| DQ402950.1 | ||||
| M16229.1 | AAA39509.1 |
More Information
Set Membership: |
|

Genomic Map