Assay Details
Target Gene Details
Entrez Gene ID: | 57370 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 3 |
Gene Aliases: |
9530061M23Rik, AA104562, AW125175, ESTM26, ESTM6, R74981 |
Location: |
Chr.1:171270328-171276895 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt3 | NM_020579.2 | NP_065604.2 | ||
| AF142671.1 | AAF22221.1 | |||
| AK155471.1 | BAE33282.1 | |||
| BC013619.1 | AAH13619.1 | |||
| BU703242.1 |
More Information
Set Membership: |
|

Genomic Map