Assay Details
Target Gene Details
Entrez Gene ID: | 66459 |
Gene Name: | Pigy upstream reading frame |
Gene Aliases: |
2610022G08Rik, AI847956, Prey |
Location: |
Chr.6:57684739-57692078 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pyurf | NM_025574.3 | 2 | 770 | NP_079850.1 |
| AK003713.1 | BAB22953.1 | |||
| AK008978.1 | ||||
| AK011515.1 | 2 | 707 | ||
| AK032381.1 | 2 | 763 | BAC27846.1 | |
| BC029232.1 | AAH29232.1 |

Genomic Map