Assay Details
Target Gene Details
Entrez Gene ID: | 72041 |
Gene Name: | alkB, alkylation repair homolog 4 (E. coli) |
Gene Aliases: |
2010004B12Rik, Abj4 |
Location: |
Chr.5:136136145-136141802 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alkbh4 | NM_028070.1 | 3 | 932 | NP_082346.1 |
| AK008083.1 | 3 | 994 | BAB25449.1 | |
| BC022729.1 | 3 | 932 | AAH22729.1 |
More Information
Set Membership: |
|

Genomic Map