Assay Details
Target Gene Details
Entrez Gene ID: | 19668 |
Gene Name: | recombination signal binding protein for immunoglobulin kappa J region-like |
Gene Aliases: |
RBP-L, Rbpsuhl |
Location: |
Chr.2:164403056-164415448 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbpjl | NM_009036.1 | NP_033062.1 | ||
| AK052449.1 | ||||
| AK131814.1 | BAE20815.1 | |||
| AK131832.1 | ||||
| BC152311.1 | ||||
| Y10926.1 | CAA71856.1 |
More Information
Set Membership: |
|

Genomic Map