Assay Details
Target Gene Details
Entrez Gene ID: | 98496 |
Gene Name: | phosphotyrosine interaction domain containing 1 |
Gene Aliases: |
5033414K04Rik, NYGGF4 |
Location: |
Chr.1:84036293-84284675 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pid1 | NM_001003948.2 | NP_001003948.2 | ||
| AK035542.1 | ||||
| AK083256.1 | ||||
| AK150971.1 | BAE30002.1 | |||
| AK151267.1 | BAE30254.1 | |||
| BC054112.1 | AAH54112.1 | |||
| BC080290.1 | AAH80290.1 |
More Information
Set Membership: |
|

Genomic Map