Assay Details
Target Gene Details
Entrez Gene ID: | 244218 |
Gene Name: | cardiotrophin 2 |
Gene Aliases: |
Gm494, NP |
Location: |
Chr.7:127717816-127725616 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctf2 | NM_198858.1 | NP_942155.1 | ||
| AB125661.1 | BAD01485.1 | |||
| AY363390.1 | AAR17733.1 | |||
| BC120763.1 | ||||
| BC120765.1 |
More Information
Set Membership: |
|

Genomic Map