Assay Details
Target Gene Details
Entrez Gene ID: | 26420 |
Gene Name: | mitogen-activated protein kinase 9 |
Gene Aliases: |
AI851083, JNK2, Prkm9, p54aSAPK |
Location: |
Chr.11:49846751-49886421 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk9 | NM_001163671.1 | NP_001157143.1 | ||
| NM_001163672.1 | NP_001157144.1 | |||
| NM_016961.3 | NP_058657.1 | |||
| NM_207692.2 | NP_997575.2 | |||
| AB005664.1 | BAA85876.1 | |||
| AF052466.1 | AAD22576.1 | |||
| AF052467.1 | AAD22577.1 | |||
| AF052468.1 | AAD22578.1 | |||
| AF052469.1 | AAD22579.1 | |||
| AK031959.1 | BAC27623.1 | |||
| AK043169.1 | BAE43319.1 | |||
| AK090057.1 | ||||
| AK145083.1 | BAE26227.1 | |||
| BC028341.1 | AAH28341.1 | |||
| CX235052.1 |
More Information
Set Membership: |
|

Genomic Map