Assay Details
Target Gene Details
Entrez Gene ID: | 21937 |
Gene Name: | tumor necrosis factor receptor superfamily, member 1a |
Gene Aliases: |
CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI, TNFRp55, TNFalpha-R1, Tnfr-2, Tnfr1, p55, p55-R |
Location: |
Chr.6:125349723-125362483 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf1a | NM_011609.4 | NP_035739.2 | ||
| AK154388.1 | BAE32554.1 | |||
| AK159989.1 | BAE35539.1 | |||
| AK211886.1 | ||||
| AY541589.1 | AAS48428.1 | |||
| AY541590.1 | AAS48429.1 | |||
| BC004599.1 | AAH04599.1 | |||
| BC052675.1 | AAH52675.1 | |||
| BC096429.1 | AAH96429.1 | |||
| CK624043.1 | ||||
| L26349.1 | AAA59361.1 | |||
| M59377.1 | AAA40464.1 | |||
| M60468.1 | AAA39751.1 | |||
| X57796.1 | CAA40936.1 | |||
| X59238.1 | CAA41922.1 |
More Information
Set Membership: |
|

Genomic Map