Assay Details
Target Gene Details
Entrez Gene ID: | 225471 |
Gene Name: | toll-like receptor adaptor molecule 2 |
Gene Aliases: |
B430113A10, TICAM-2, TRAM, Tirp, Trif |
Location: |
Chr.18:46558231-46574533 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ticam2 | NM_173394.3 | NP_775570.1 | ||
| AB100441.1 | BAC98398.1 | |||
| AK080873.1 | BAC38055.1 | |||
| AK134103.1 | BAE22014.1 | |||
| AK151885.1 | BAE30771.1 | |||
| AY275837.1 | AAP81749.1 | |||
| BC099933.1 | AAH99933.1 |
More Information
Set Membership: |
|

Genomic Map