Assay Details
Target Gene Details
Entrez Gene ID: | 56746 |
Gene Name: | testis expressed gene 101 |
Gene Aliases: |
1700008H15Rik, AI429076, TES101RP |
Location: |
Chr.7:24668012-24672050 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tex101 | NM_019981.2 | 6 | 960 | NP_064365.1 |
| AB022914.1 | 6 | 958 | BAA90265.1 | |
| AK005769.1 | 6 | 954 | BAB24228.1 | |
| BC048475.1 | 6 | 951 | AAH48475.1 |

Genomic Map