Assay Details
Target Gene Details
Entrez Gene ID: | 75584 |
Gene Name: | RIKEN cDNA 2310030A07 gene |
Gene Aliases: |
- |
Location: |
Chr.1:150320681-150346771 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310030A07Rik | NR_040603.1 | |||
| AK009534.1 |

Genomic Map