Assay Details
Target Gene Details
Entrez Gene ID: | 18511 |
Gene Name: | paired box 9 |
Gene Aliases: |
Pax-9 |
Location: |
Chr.12:56690898-56712824 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pax9 | NM_011041.2 | NP_035171.1 | ||
| AK034985.1 | BAC28903.1 | |||
| AK132404.1 | BAE21149.1 | |||
| BC005794.1 | AAH05794.1 | |||
| X84000.1 | CAA58824.1 |
More Information
Set Membership: |
|

Genomic Map