Assay Details
Target Gene Details
Entrez Gene ID: | 68115 |
Gene Name: | RIKEN cDNA 9430016H08 gene |
Gene Aliases: |
AA409999, C78988 |
Location: |
Chr.1:57406548-57415958 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9430016H08Rik | NM_001081181.2 | NP_001074650.1 | ||
| AK007831.1 | ||||
| AK079116.1 | BAC37547.1 | |||
| AK165130.1 | 1 | 2691 | ||
| BC024945.1 | AAH24945.1 |
More Information
Set Membership: |
|

Genomic Map